missing translation for 'onlineSavingsMsg'
Learn More

Setiptiline maleate, MedChemExpress

Product Code. 16490488
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490488 5 mg 5mg
16487806 100 mg 100mg
16453957 50 mg 50mg
16408038 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490488 Supplier Medchem Express Supplier No. HY32329A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Setiptiline maleate (MO-8282 maleate) is a serotonin receptor antagonist.

Setiptiline maleate is a tetracyclic antidepressant (TeCA) which acts as a noradrenergic and specific serotonergic antidepressant (NaSSA). Setiptiline maleate acts as a norepinephrine reuptake inhibitor, α2-adrenergic receptor antagonist, and serotonin receptor antagonist, likely at the 5-HT2A, 5-HT2C, and/or 5-HT3 subtypes, as well as an H1 receptor inverse agonist/antihistamine.

Specifications

Chemical Name or Material Setiptiline maleate
CAS 85650-57-3
Color White
Purity Grade Notes Research
Molecular Formula C23H23NO4
Quantity 5 mg
Synonym MO-8282
Solubility Information DMSO : 100 mg/mL (264.95 mM; Need ultrasonic)
SMILES CN1CC(C(C=CC=C2)=C2C3)=C(CC1)C4=C3C=CC=C4.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 377.43
Formula Weight 377.43
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.