Learn More
BD Biosciences™ Setup Beads (CE/IVD)
Product Code. 15886379 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
Each
Product Code. Quantity Unit Size
15886379 25 Tests Each
1 options
Product Code. 15886379 Supplier BD Biosciences™ Supplier No. 658620
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Intended to set voltages appropriate for the BD multicolor tube assay when used with a suitably equipped BD™ flow cytometer and software designated for in vitro diagnostic use.
  • Set voltages appropriate for the BD multicolor tube assay.
  • Fluorescent particles that are used to set cytometer detector photomultiplier tube voltages (PMTVs) for the BD multicolor tube assay.
  • PMTVs are manually adjusted to place the BD OneFlow Setup beads at their lot specific median fluorescence intensity (MFI) target ranges for all fluorescence parameters.
  • Lysed washed blood (LWB) is used to set cytometer FSC and SSC voltages to a target value range.
  • The detector settings are then saved as Application Settings.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage One vial of BD OneFlow Setup beads, Monthly MFI target range card, Daily MFI target range card
No. of Tests 25 Tests
Quantity 25 Tests
Stability After dilution, the beads are stable for 1 hour at 18%deg;C to 25°C, and for 8 hours at 2°C to 8°C
Product Type Fluorescent Polystyrene Beads