missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ S210 SevenCompact™ pH, mV/ORP-Kit

Product Code. 15304278 Shop All Mettler Toledo Products
Change view
Click to view available options
Resolution (mV):
0.1/1mV
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Resolution (mV) unitSize
15304278 0.1/1mV Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15304278 Supplier Mettler Toledo™ Supplier No. 30130863

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This well-designed meter is the perfect choice for a wide range of applications without overstretching your budget. Mettler Toledo™ S210 SevenCompact™ pH, mV/ORP-Kit can also measure ORP.

  • Includes electrode holder
  • UPlace™ movement helps bring your electrode into the best position for your sample
  • Fast measurements and reduced sample tipping risk
  • Reduced sensor damage risk
  • Choice of viewing all measurement information on the display or focusing on the essentials with the uFocus™ view
  • Switching between the different display layouts requires only a single keypress
  • Color display with well designed icons
  • Menu settings in 10 languages
  • Security and high reproducibility thanks to Intelligent Sensor Management (ISM™)
  • Professional calibration support
  • Comprehensive service package
  • Integrated USB and RS232 interfaces
  • Includes InLab™ Expert Pro-ISM, guide to pH measurements and 2 buffer sachets for pH 4.01, 7.00, 9.21, and 10.0
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Battery Type 9 to 12 V
Calibration Points Max. 5 points, 8 pre-defined and 1 user-defined buffer group
Description SevenCompact pH meter S210-Std-Kit
Detection Range -2 to 20°C
Channels 1
Display Type TFT Color, 4.34 in.
Range (mV) -2000 to 2000.0
Temperature Range (Metric) -30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC)
For Use With (Application) Measurements of pH, mV/ORP
Includes InLab™ Expert Pro-ISM, guide to pH measurements and 2 buffer sachets for pH 4.01, 7.00, 9.21, and 10.0
Languages Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 1.000 measurements
Model pH bench meter SC S210-K
Power Supply 9 to 12V/10W
Product Type pH/mV/ORP Meter
Resolution (mV) 0.1/1mV
Temperature Accuracy ±0.1°C
Electrical Requirements 9 to 12V, 10W
Voltage 9/12 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.