missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Pole for EasyPlace Electrode Arm

Product Code. 17511026 Shop All Mettler Toledo Products
Click to view available options
Type:
Extended Pole, EasyPlace, 50 cm
Standard Pole, EasyPlace, 30 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17511026

Brand: Mettler Toledo™ 30671577

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pole for the EasyPlace electrode arm.

  • Accessory
TRUSTED_SUSTAINABILITY

Specifications

Type Standard Pole, EasyPlace, 30 cm
Size 30 cm
For Use With (Equipment) EasyPlace
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.