Learn More
SGE™ ConnecTite™ Liners for Thermo Scientific™ GC
Product Code. 15279023
Change view
Click to view available options
Product Type:
Connectite Bottom Hole Liner
Connectite Standard Liner
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15279023 Connectite Bottom Hole Liner Pack of 5
15239023 Connectite Standard Liner Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15279023 Supplier Trajan Supplier No. 092336
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for use with Thermo Scientific TRACE™ and FOCUS™ GCs
  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 8mm OD, 5mm ID; 105mm length
Specifications
Product Type Connectite Bottom Hole Liner
Quantity 5/Pk.
Length (Metric) 10.5 cm
Diameter (Metric) Inner 0.5 cm
Diameter (Metric) Outer 0.8 cm
For Use With (Equipment) Thermo Scientific GC Trace and Focus