Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Improves laboratory workflow
89.60 € - 357.00 €
Product Code | Brand | Product Type | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
12927586
|
SGE™
2910020 |
eVol Analytical Syringe | 5 μL | Ea. |
96.50 €
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12997586
|
SGE™
2910024 |
eVol Analytical Syringe | 500 μL | Ea. |
96.50 €
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12947586
|
SGE™
2910022 |
eVol Analytical 25 ga | 50 μL | Ea. |
96.50 €
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12907596
|
SGE™
2910025 |
eVol™ Analytical Syringe, No Needle | 20 to 500 μL | Ea. |
89.60 €
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12927596
|
SGE™
2910320 |
eVol Analytical Syringe | 5 μL | 3/Pk. |
259.00 €
Pack of 3 |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12937596
|
SGE™
2910322 |
eVol Analytical Syringe | 50 μL | 3/Pk. |
357.00 €
Pack of 3 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12957596
|
SGE™
2910324 |
eVol Analytical Syringe | 500 μL | 3/Pk. |
259.00 €
Pack of 3 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
- Digitally controlled positive displacement dispensing system
- Programmable—enables user-independent liquid handling procedures
- Easily calibrated to ensure accurate dispensing
- Stores calibration factors for up to ten XCHANGE™ syringes
- Touch Wheel user interface
- Full color screen
Applications:
- Preparation of calibration standards
- Addition of internal standards
- Precise dispensing of non-aqueous liquids
- Sample dilution