missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Replacement Parts for Manual Syringes

Product Code. 15258573
Change view
Click to view available options
For Use With (Equipment):
Luer Lock Syringe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15258573 Luer Lock Syringe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15258573 Supplier Trajan™ Supplier No. 031902
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement parts for removable Luer Lock syringes

  • Replacement Luer Lock fitting
  • Contains 1 Kel-F cone and 1 threaded metal adaptor

Specifications

Includes 1 Kel-F luer cone and 1 threaded metal adaptor
Product Type Luer Lock Fitting
For Use With (Equipment) Luer Lock Syringe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.