Learn More
Trajan™ Terry Tool Tubing Cutter
Product Code. 10547732
Change view
Click to view available options
Diameter (English) Outer:
1/16 in.
Unit Size:
Each
Product Code. Diameter (English) Outer Unit Size
10547732 1/16 in. Each
1 options
Product Code. 10547732 Supplier Trajan™ Supplier No. 082780
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Terry Tool Tubing Cutter for accurate cutting of stainless steel tubing
  • Accurate cutting of stainless steel tubing
  • Burr-free tubing ends
Specifications
Type Tubing Cutter
Diameter (English) Outer 1/16 in.
Diameter (Metric) Outer 1.59 mm
For Use With (Application) Cutting Stainless Steel Tubing
Quantity 1/Pk.