Learn More
Manutan Shackles, 2.75T large bow, high tensile steel
Product Code. 12852758
Encompass_Preferred
Product Code. 12852758
Brand: Manutan A340016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • High tensile steel
  • Dimensions : d - 22mm ; D - 25mm ; w - 41mm ; 2r - 56mm ; s - 98mm ; e - 50mm