Learn More
Manutan Shackles, 2 8.5T SWL D, galvanized alloy steel
Product Code. 12868788
Encompass_Preferred
Product Code. 12868788 Supplier Manutan Supplier No. A340009
Estimated Shipment: 22-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • Galvanised alloy steel
  • Dimensions : d - 25mm ; D - 28mm ; w - 43mm ; s - 85mm ; e - 56mm