Learn More
Memmert™ Shaking Device for Waterbaths With Support Frame
Product Code. 17150335 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
WTB15
WTB24
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17150335 WTB15 Each
17150365 WTB24 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17150335 Supplier Memmert™ Supplier No. WTB15SV1524
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Shaking device with support frame without gable cover & support basket (mandatory)
  • Requires gable cover
  • Voltage 230 V, 50/60 Hz
  • Order the support basket separately
  • Includes support frame, shaking frequency 35 - 160 strokes per minute (horizontal back/forth movement)
Specifications
For Use With (Equipment) WTB15
Type Shaking device for waterbaths incl. support frame
Speed 35-160 rpm