Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II, White
Product Code. 12700928 Shop All Epredia Products
missing translation for 'orderingAttributeHoverText'
Color:
Lilac
White
missing translation for 'unitSize'
Each
Pack of 500
Product Code. 12700928
missing translation for 'mfr': Epredia™ 1001097
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Hold biopsy specimens during processing and embedding with the special design of Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II.
  • Acetal polymer construction
  • Locking lid attached by two hinges
  • Cassette writing face is at a 45° angle
  • Pore style is small square
  • Variety of colors available
Ordering Alerts:
  • Supplied in boxes of 500 cassettes with attached lids
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Square Pore
Writing Face Angle 45°
Color White
Material Acetal Polymer
Product Type Biopsy Cassette