missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Bluing Reagent

Product Code. 10381775 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4L
2 x 1L
Unit Size:
4 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10381775 2 x 1L Each
10127998 4L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10381775 Supplier Epredia™ Supplier No. 6769001
  • ONLINE EXCLUSIVE PRICE
    51.95 €
    40.37 € / Pack of 1
    Save 11.58 €
    22% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.

  • Ready-to-use
  • Thoroughly pH-controlled for completely effective bluing
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 x 1L
Product Type Bluing Reagent
compliance-icons compliance-icons compliance-icons
Product Identifier
  • shandon bluing reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P235-Keep cool.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium bicarbonate,lithium carbonate,methyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.