Learn More
Epredia™ Cytoblock™ Cell Block Preparation System
Product Code. 10066588 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cell Preparation System
Replacement Reagent
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10066588 Cell Preparation System Each
10446640 Replacement Reagent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10066588 Supplier Epredia™ Supplier No. 7401150
  • ONLINE EXCLUSIVE PRICE
    130.00 €
    93.70 € / Pack of 1
    Save 36.30 €
    27% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cell Preparation System