Learn More
Epredia™ Double Cytofunnel™
Product Code. 12086689 Shop All Epredia Products
Change view
Click to view available options
Quantity:
25 Pk.
500/Cs.
Unit Size:
Pack of 25
Pack of 500
Product Code. Quantity unitSize
12086689 500/Cs. Pack of 500
12016679 25 Pk. Pack of 25
2 options
Product Code. 12086689
Brand: Epredia™ 1102547
Only null left
Add to basket
Add to basket
Optimize cell yield with the Epredia™ Double Cytofunnel™.
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Reduces the risk of exposure to pathogenic samples
  • Reduces preparation time
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Epredia™ Double Cytofunnel™ REQUIRES Epredia™ Cytoclips™
  • Decreases the possibility of cross-contamination
  • Features funnel markings corresponding to slide markings for simple identification
Includes:
  • Pre-attached filter card
  • Closure caps
  • Easy-to-use operational illustrations
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Single-use
Quantity 500/Cs.
Type Double Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.05 to 0.25 mL