Learn More
Epredia™ EZ Double Cytofunnel™
Product Code. 12076689 Shop All Epredia Products
Change view
Click to view available options
Quantity:
200/Cs.
40 Pk.
Unit Size:
Pack of 200
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12076689 200/Cs. Pack of 200
11911788 40 Pk. Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12076689 Supplier Epredia™ Supplier No. A78710021
  • ONLINE EXCLUSIVE PRICE
    461.00 €
    357.72 € / Pack of 200
    Save 103.28 €
    22% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Double the number of sample presentations per run with the Epredia™ EZ Double Cytofunnel™.
  • Disposable sample chambers with white filter cards and caps
  • Funnel markings corresponding to slide markings for simple identification
  • Pre-attached filter card
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Single-use
Quantity 200/Cs.
Type EZ Double Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.05 to 0.25 mL