Learn More
Epredia™ Shandon™ Finesse™ Fine Orienting Head
Product Code. 12658046 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Fine Orienting Head
Unit Size:
Each
Product Code. Product Type Unit Size
12658046 Fine Orienting Head Each
1 options
Product Code. 12658046 Supplier Epredia™ Supplier No. 77510166
  • ONLINE EXCLUSIVE PRICE
    488.00 €
    394.48 € / Pack of 1
    Save 93.52 €
    19% Off
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for the Epredia™ Shandon™ Finesse ME/E Series only, the Epredia™ Finesse Fine Orienting Head is of excellent quality.
  • Allows x-y orientation of the specimen
  • Supplied with the Finesse ME+ and E+ model microtomes only
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fine Orienting Head