Learn More
Epredia™ Small Charcoal Filter
Product Code. 12623106 Shop All Epredia Products
Change view
Click to view available options
Quantity:
6 Pk.
Unit Size:
Each
Product Code. Quantity Unit Size
12623106 6 Pk. Each
1 options
Product Code. 12623106 Supplier Epredia™ Supplier No. 9990610
  • ONLINE EXCLUSIVE PRICE
    219.00 €
    178.29 € / Pack of 1
    Save 40.71 €
    18% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Epredia™ Hyperclean Pathology Workstations.
Accessories:
  • Charcoal Filter
  • Fine particle Filters
  • Formaldehyde Neutralizing (Potassium Permanganate) Filter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Charcoal Filter
Description Size: 29 cm x 15 cm x 3.3 cm
For Use With (Equipment) Epredia Hyperclean Pathology Workstations, Models 1, 20, 22, 36, 60
Product Line Hyperclean
Quantity 6 Pk.