missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Shandon™ Paraffin

Product Code. 15189903 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Paraffin
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15189903 Paraffin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15189903 Supplier Thermo Scientific™ Supplier No. 502004

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Achieve thorough tissue infiltration and provide maximum support during microtomy with high-quality Thermo Scientific™ Shandon™ Paraffin.

  • Quality blend of paraffin, polymers and plastics (to varying degrees, based upon type)
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 56°C
Product Type Paraffin
Quantity 8 x 1kg
For Use With (Application) Enhanced infiltration and sectioning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.