missing translation for 'onlineSavingsMsg'
Learn More

Shanzhiside Methyl Ester, TRC

Product Code. 30411009
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411009 2.5 mg 2.5mg
30420975 5 mg -
30410589 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411009 Supplier Toronto Research Chemicals Supplier No. TRCS2823902.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64421-28-9
Molecular Formula C17 H26 O11
Molecular Weight (g/mol) 406.38
Synonym Cyclopenta[c]pyran-4-carboxylic acid, 1-(β-D-glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-5,7-dihydroxy-7-methyl-, methyl ester, (1S,4aS,5R,7S,7aS)-, Cyclopenta[c]pyran-4-carboxylic acid, 1-(β-D-glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-5,7-dihydroxy-7-methyl-, methyl ester, [1S-(1α,4aα,5α,7α,7aα)]-, Sanshiside methyl ester, Shanzhiside methyl ester, Shanzhizide methyl ester, Shanziside methyl ester
IUPAC Name methyl (1S,4aS,5R,7S,7aS)-5,7-dihydroxy-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4a,5,6,7a-tetrahydro-1H-cyclopenta[c]pyran-4-carboxylate
SMILES COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3[C@@H]1[C@H](O)C[C@]3(C)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H26O11/c1-17(24)3-7(19)9-6(14(23)25-2)5-26-15(10(9)17)28-16-13(22)12(21)11(20)8(4-18)27-16/h5,7-13,15-16,18-22,24H,3-4H2,1-2H3/t7-,8-,9+,10-,11-,12+,13-,15+,16+,17+/m1/s1
Formula Weight 406.1475
Chemical Name or Material Shanzhiside methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.