Learn More
Honeywell™ SHARPFLEX Liner Gloves
Description
- Knitted glove in composite and Kevlar yarn
- The knitting technique provides the glove with excellent fit and high dexterity
- The composite yarn is reliable and safe protection
- The Kelvar yarn assure a resistance to the contact heat and cut risks
- Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
- Being Ambidexterous increases the lifetime of the glove
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards, EN 407-04 Thermical hazards |
| Description | 13 gauge, Dupont™ Kevlar™ aramid fibers and composite fiber |
| Color | Green, Yellow |
| Hand | Ambidextrous |
| Material | Polyamide |
| Size | 9 |
| Length (Metric) | 25 cm |