Learn More
Honeywell™ SHARPFLEX Liner Gloves
Product Code. p-7159530
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Product Code. 15876725
Brand: Honeywell™ 223252207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ SHARPFLEX Liner Gloves for handling heavy parts with high cut risks in a dry environment.
  • Knitted glove in composite and Kevlar yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The Kelvar yarn assure a resistance to the contact heat and cut risks
  • Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
  • Being Ambidexterous increases the lifetime of the glove
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards, EN 407-04 Thermical hazards
Description 13 gauge, Dupont™ Kevlar™ aramid fibers and composite fiber
Color Green, Yellow
Hand Ambidextrous
Material Polyamide
Size 7
Length (Metric) 25 cm