Learn More
Honeywell™ SHARPFLEX NIT Gloves
Description
- Knitted glove in composite and para-aramid yarn
- The knitting technique provides the glove with excellent fit and high dexterity
- The composite yarn is reliable and safe protection
- The para-aramid yarn assure a resistance to the contact heat and cut risks
- Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
- Black nitrile foam coating on palm
- Nitrile coating provides good protection against oil and grease
- The foam coating makes the glove more flexible than standard nitrile
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards |
| Description | Para-aramid and composite, 13 gauge |
| Color | Green, Yellow, Black |
| Material | Nitrile, Polyamide |
| Size | 7 |
| Length (Metric) | 25 cm |