missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ SHARPFLEX NIT Gloves

Product Code. 15886735
Change view
Click to view available options
Size:
7
8
9
10
11
Packungsgröße:
1 pair
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packungsgröße
15886735 8 1 pair
15876735 7 1 pair
15896735 9 1 pair
15806745 10 1 pair
15816745 11 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 15886735 Lieferant Honeywell™ Lieferanten-Nr. 223252408
  • ONLINE EXCLUSIVE PRICE
    13.42 €
    9.51 € / 1 pairs
    Save 3.91 €
    29% Off
In Stock
Zum Warenkorb hinzufügen
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell™ SHARPFLEX NIT Gloves for handling heavy parts with high cut risks in a dry or slightly greasy environment.

  • Knitted glove in composite and para-aramid yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The para-aramid yarn assure a resistance to the contact heat and cut risks
  • Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
  • Black nitrile foam coating on palm
  • Nitrile coating provides good protection against oil and grease
  • The foam coating makes the glove more flexible than standard nitrile
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards
Description Para-aramid and composite, 13 gauge
Color Green, Yellow, Black
Material Nitrile, Polyamide
Size 8
Length (Metric) 25 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.