missing translation for 'onlineSavingsMsg'
Learn More

Sharpsafe™ Polypropylene Biohazard Sharps Container

Product Code. 11930368
Change view
Click to view available options
Capacity (Metric):
4 L
7 L
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11930368 7 L Pack of 50
11920368 4 L Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11930368 Supplier Sharpsafe™ Supplier No. SHA4110

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use the Sharpsafe™ Polypropylene Biohazard Sharps Container facilitate the safe management of wastes from healthcare activities.

  • Polypropylene
  • 50Pack
  • Yellow

Specifications

Capacity (Metric) 7 L
Color Yellow
Height (Metric) 382 mm
Length (Metric) 175 mm
Quantity 50 Pk.
Width (Metric) 175 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.