missing translation for 'onlineSavingsMsg'
Learn More

Sharpsafe™ Polypropylene Medicinal Sharps Container

Product Code. 12943535
Change view
Click to view available options
Capacity (Metric):
1.0 L
2.0 L
4 L
7 L
9 L
13 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12943535 13 L Each
12983525 4 L Each
12903535 7 L Each
12923535 9 L Each
12943525 1.0 L Each
12963525 2.0 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12943535 Supplier Sharpsafe™ Supplier No. 41152430

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for use in public access areas such as A&E and Out-patients. Sharpsafe™ Polypropylene Medicinal Sharps Container allows easy disposal of sharps but provides added security for both the user and the general public. The Protected Access range also discourages the use of the sharps container for non sharps waste, avoiding unnecessary waste disposal costs.

  • Easy disposal of sharps
  • Protected Access range 
  • Polypropylene

Specifications

Capacity (Metric) 13 L
Certifications/Compliance BS 7320, AFNOR NX30-500, UN 3291
Closure Type Lid
Closure Color Yellow
Color Yellow
Model 4115
Quantity 1 Ea.
Includes Closure
For Use With (Application) Designed for use in public access areas such as A&E and out-patients
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.