missing translation for 'onlineSavingsMsg'
Learn More

Sharpsguard™ yellow 22 Multi-Purpose 17.86L Sharps Container with Pail Handle

Ideal for theaters, laboratories and sluice rooms with high volumes of hazardous waste. Sharpsguard™ yellow 22 Multi-Purpose 17.86L Sharps Container with Pail Handle is for disposal of sharps that have been contaminated with medicinal products, but not cytotoxic or cytostatic.

Brand:  Daniels Healthcare DD475YL

Code : 02

Additional Details : Weight : 9.36000kg

Product Code. 12963495

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Container for disposal by incineration
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • Wide aperture allows economical and convenient disposal of large items
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications

Specifications

Polypropylene
BS7320:1990, UN3291, AFNOR, NFX 30-500
Yellow
330 mm
Autoclavable
17.86 L
Lid
345 mm
148 x 78 mm (L x W)
Ideal for theatres, laboratories and sluice rooms with high volumes of healthcare waste
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers