missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sheep anti-Bovine IgG1 Secondary Antibody, HRP

Product Code. 11539200
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11539200 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11539200 Supplier Invitrogen™ Supplier No. PA184658
  • ONLINE EXCLUSIVE PRICE
    538.00 €
    478.82 € / 1mg
    Save 59.18 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Secondary Antibody

Sheep anti Bovine IgG1 polyclonal antibody recognizes bovine IgG1. No cross-reactivity with other bovine immunoglobulin classes is seen in immunoelectrophoresis.This product may cross-react with IgG1 from other species.

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine IgG1
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.05% ProClin 300
Host Species Sheep
Immunogen Purified bovine IgG1.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.