missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sheep anti-Rat IgG (H+L) Secondary Antibody, Biotin

Product Code. 11565953
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11565953 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11565953 Supplier Invitrogen™ Supplier No. PA128641
  • ONLINE EXCLUSIVE PRICE
    292.00 €
    259.88 € / 1mg
    Save 32.12 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Secondary Antibody

PA1-28641 detects Rat IgG from rat samples.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Spécification

Antigen Rat IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA and 0.01% sodium azide; pH 7.2
Host Species Sheep
Immunogen Rat IgG whole molecule.
Purification Method IgG fraction
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.