Learn More
Sheep IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Donkey Polyclonal Antibody
Brand:  R&D Systems VC006-250
 View more versions of this product
Product Code. 18717828
  • 2590.00 € / 250mL
Description
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Sheep IgG
250 mL (2 x 125 mL)
Secondary
Detects heavy ( gamma ) chains on sheep IgG and light chains on all sheep immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin sheep serum proteins or IgG from human, mouse or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
6 months from date of receipt, 2 to 8 degreesC as supplied.
Immunohistochemistry
HRP Polymer
Supplied as a solution in PBS with preservative.
Donkey
Affinity Purified
RUO
3500
Sheep