missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Sheep TP-1 (NP_001116871) VersaClone cDNA
Description
- Also known as: OTP; IFNT1; IFNT3; IFNT4; IFNT7; IFNT8; IFNT9; TP-07; TP-08; TP-010; IFN-tau-3; IFN-tau-7; IFN-tau-8; IFN-tau-9
- TP-1 belongs to the alpha/beta interferon family.
- It is primarily responsible for the maternal recognition of pregnancy.
- TP-1 interacts with endometrial receptors, probably type I interferon receptors, and blocks estrogen receptor expression, preventing the estrogen-induced increase in oxytocin receptor expression in the endometrium.
- It is constitutively and exclusively expressed in the mononuclear cells of the extraembryonic trophectoderm.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | TP-1 |
| Accession Number | NP_001116871 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 601 bp |
| Gene ID (Entrez) | 100144750 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Sheep |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?