missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mylar sheet for Vacuum Gel Dryers

Product Code. 12368546
Change view
Click to view available options
For Use With (Equipment):
FGD-5040 Fisherbrand Vacuum Gel Dryer System
Mylar Fisherbrand Gel Dryer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12368546 FGD-5040 Fisherbrand Vacuum Gel Dryer System Each
11784306 Mylar Fisherbrand Gel Dryer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12368546 Supplier Fisherbrand™ Supplier No. GDMS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessory for use with vacuum gel dryers

  • Clear cover for viewing of the drying process
  • Used as an accessory for Vacuum gel dryer systems

Tekniske data

Color White
For Use With (Equipment) FGD-5040 Fisherbrand Vacuum Gel Dryer System
Type Sheet
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.