missing translation for 'onlineSavingsMsg'
Learn More

Sheet Steel Oxygen Cylinder Trolleys

Product Code. 12945961
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W x H):
460 x 410 x 1100 mm
470 x 480 x 1100 mm
missing translation for 'unitSize'
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12945961 470 x 480 x 1100 mm Each
12935961 460 x 410 x 1100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 12945961 missing translation for 'mfr' Brand Not Defined missing translation for 'supplierNo' TS180L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The tubular frame has a sheet steel base and retaining straps to hold the cylinder in position. Each trolley is fitted with anti-static metal centred wheels and front 100mm non-marking corner buffers.

Specifications

Type Oxygen Cylinder Trolley
Material Sheet Steel
Dimensions (L x W x H) 470 x 480 x 1100 mm
For Use With (Application) Ideal for use in hospitals
For Use With (Equipment) Carrying oxygen cylinders
Height (Metric) 1100 mm
Length (Metric) 470 mm
Weight (Metric) 14 kg
Width (Metric) 480 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.