Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Product Code. 11850543 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Boxes (100/4 Racks)
Boxes (100/5 Racks)
Boxes (120/6 Racks)
Boxes (125/5 Racks)
Boxes (140/7 Racks)
Boxes (150/6 Racks)
Boxes (175/7 Racks)
Boxes (48/4 Racks)
Boxes (60/3 Racks)
Boxes (75/3 Racks)
Boxes (80/4 Racks)
Boxes (80/5 Racks)
For Use With (Equipment):
Forma 900, Forma 7000, TSE and TSD Series (13 cu.ft.)
Forma 900, Forma 7000, TSE and TSD Series (17 cu.ft.)
Forma 900, Forma 7000, TSE and TSD Series (23 cu.ft.)
HERAfreeze HFU B Series, Revco ExF and DxF Series
HERAfreeze HFU B Series, Revco ExF and DxF Series (13 cu.ft.)
HERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu.ft.)
Thermo Scientific TDE Series (14.9 cu.ft.)
Thermo Scientific TSX Series (33.5 cu.ft.)
Thermo Scientific TSX and TDE Series (19 cu.ft.)
Thermo Scientific TSX and TDE Series (24.1 cu.ft.)
Thermo Scientific TSX and TDE Series (28.8 cu.ft.)
No. of Racks:
3
4
5
6
7
Unit Size:
Each
Product Code. Holds For Use With (Equipment) No. of Racks unitSize
11850543 Boxes (175/7 Racks) Thermo Scientific TSX Series (33.5 cu.ft.) 7 Each
12908344 Boxes (48/4 Racks) Forma 900, Forma 7000, TSE and TSD Series (13 cu.ft.) 4 Each
11329351 Boxes (48/4 Racks) HERAfreeze HFU B Series, Revco ExF and DxF Series (13 cu.ft.) 4 Each
12918344 Boxes (80/4 Racks) Forma 900, Forma 7000, TSE and TSD Series (17 cu.ft.) 4 Each
11339351 Boxes (80/4 Racks) HERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu.ft.) 4 Each
12928344 Boxes (100/5 Racks) Forma 900, Forma 7000, TSE and TSD Series (23 cu.ft.) 5 Each
11349351 Boxes (80/5 Racks) HERAfreeze HFU B Series, Revco ExF and DxF Series 5 Each
11810543 Boxes (75/3 Racks) Thermo Scientific TDE Series (14.9 cu.ft.) 3 Each
11359351 Boxes (60/3 Racks) Thermo Scientific TDE Series (14.9 cu.ft.) 3 Each
11820543 Boxes (100/4 Racks) Thermo Scientific TSX and TDE Series (19 cu.ft.) 4 Each
11369351 Boxes (80/4 Racks) Thermo Scientific TSX and TDE Series (19 cu.ft.) 4 Each
11830543 Boxes (125/5 Racks) Thermo Scientific TSX and TDE Series (24.1 cu.ft.) 5 Each
11379351 Boxes (100/5 Racks) Thermo Scientific TSX and TDE Series (24.1 cu.ft.) 5 Each
11840543 Boxes (150/6 Racks) Thermo Scientific TSX and TDE Series (28.8 cu.ft.) 6 Each
17708049 Boxes (120/6 Racks) Thermo Scientific TSX and TDE Series (28.8 cu.ft.) 6 Each
11399351 Boxes (140/7 Racks) Thermo Scientific TSX Series (33.5 cu.ft.) 7 Each
16 options
Product Code. 11850543
Brand: Thermo Scientific™ RSK700SD4
Only null left
Add to basket
Add to basket
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
TRUSTED_SUSTAINABILITY
Specifications
Cell Dividers 100
Description Shelf Kit
Holds Boxes (175/7 Racks)
Material Stainless Steel
For Use With (Equipment) Thermo Scientific TSX Series (33.5 cu.ft.)
No. of Racks 7
Product Type Shelf Kit
Shelves 4 Tiers