missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Shelf

Product Code. 15876392 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
329 x 376 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15876392 329 x 376 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15876392
  • ONLINE EXCLUSIVE PRICE
    148.00 €
    133.00 € / Pack of 1
    Save 15.00 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Material: Stainless Steel; For Use With: Isotemp gravity Incubator, 2.6 cu. ft.

  • Material: Stainless steel
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Isotemp gravity incubator, 2.6 cu. ft.
Material Stainless Steel
Type Shelf
Dimensions (L x W) 329 x 376 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.