missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Shelves for FreeZone™ Freeze Dryers

Product Code. 15777807 Shop All Labconco Corporation Products
Change view
Click to view available options
Size:
Short
Tall
No. of Shelves:
3
5
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size No. of Shelves Unit Size
15777807 Tall 5 Each
15767807 Short 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15777807 Supplier Labconco™ Supplier No. 7317200
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.

  • Removable, 10 in. diameter aluminum shelves with 1/4 in. rim
  • Shelves slide into stainless steel support
  • Includes 3 rubber feet
  • Design allows for flexible shelf height spacing
  • Shelf can be placed in freezer to prefreeze sample for quick loading
  • Designed to fit inside Drying Chambers (sold separately)
  • Also available: Heated shelves that include ambient to +40°C (+104°F) variable heat control on models manufactured 2016 and earlier and up to +60°C (+140°F) on models 2017 and later (contact your Fisher Customer Service Representative for details).

Specifications

Type Product Shelves
Capacity (Metric) 250 L
No. of Shelves 5
Material Stainless Steel
Size Tall
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment) FreeZone™ Drying Chambers
Includes Five aluminum shelves with 1/4 in. rims, one stainless steel support, and three rubber feet
Width (English) 10.4 in.
Height (English) 17.5 in.
Height (Metric) 44.45 cm
Length (English) 10.9 in.
Length (Metric) 27.7 cm
Shipping Weight (English) 7 lb.
Weight (English) 5.5 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.