missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Shielded GSIOC Cable

Product Code. 17173588 Shop All Gilson Products
Click to view available options
For Use With (Equipment):
GSIOC Communicating Instruments
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17173588

Brand: Gilson™ 36078143

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This Shielded GSIOC Cable is for use with GSIOC communicating instruments.

  • Length of 76.2 cm (30 in.)
  • For use with GSIOC communicating instruments

Specifications

For Use With (Equipment) GSIOC Communicating Instruments
Type GSIOC Cable
Length (Metric) 76.2 cm
Length (English) 30 in.
Product Line ALH Product Line
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.