missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Shimadzu GC-2010 Liners

Product Code. 15239013
Change view
Click to view available options
Product Type:
Inlet liners PTV 3 baffle
Straight LVI Liner with Wool
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15239013 Straight LVI Liner with Wool Pack of 5
15219013 Inlet liners PTV 3 baffle Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15239013 Supplier Trajan Scientific and Medical™ Supplier No. 092292
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Provides excellent analysis for a wide range of compounds.

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Shimadzu instruments

Specifications

Product Type Straight LVI Liner with Wool
Description GC-2010 Liners
Diameter (Metric) Inner 1.5 mm
Diameter (Metric) Outer 3.1 mm
For Use With (Equipment) Shimadzu Instruments
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.