Learn More
Dräger™ Short-Term Detector Tubes: Chlorine
Product Code. 10178231
Change view
Click to view available options
Measurement Range:
0.2 to 30
0.3 to 5
50 to 500
Tube Type:
Chlorine 0.2/a
Chlorine 0.3/b2
Chlorine 50/a
Humidity Range:
Less than15mg/L
Less than40mg/L
Not Applicable
No. of Strokes:
1
10/1
20
Tamaño de la unidad:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Measurement Range Tube Type Humidity Range No. of Strokes unitSize
10178231 0.2 to 30 Chlorine 0.2/a Not Applicable 10/1 Pack of 10
10558292 0.3 to 5 Chlorine 0.3/b2 Less than15mg/L 20 Pack of 10
15778659 50 to 500 Chlorine 50/a Less than40mg/L 1 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10178231 Proveedor Dräger™ N.º de proveedor CH24301
Este artículo no se puede devolver. Vea la política de devoluciones
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Información de órdenes
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Especificaciones
Components Measured Chlorine
Humidity Range Not Applicable
Measurement Range 0.2 to 30
No. of Strokes 10/1
Quantity 10
Response Time 3 min./20 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Chlorine 0.2/a
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.