missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide

Product Code. 15365442
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide
Hydrogen Sulfide 0.2%/a
Hydrogen Sulfide 0.2/b
Hydrogen Sulfide 2%/A
Hydrogen Sulfide 2/a
Hydrogen Sulfide 2/b
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
Measurement Range:
0.2 to 5ppm
0.2 to 6ppm
0.2 to 7 vol.%
2 to 200ppm
2 to 40 vol.%
2 to 60ppm
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Unit Size
15365442 Hydrogen Sulfide 2/a 2 to 200ppm Pack of 10
10473172 Hydrogen Sulfide 2/b 2 to 60ppm Pack of 10
10347892 Hydrogen Sulfide 0.2 to 5ppm Pack of 10
10388421 Hydrogen Sulfide, Sulfur Dioxide 0.2%/A 0.2 to 7 vol.% Each
10153671 Hydrogen Sulfide 0.2%/a 0.2 to 7 vol.% Pack of 10
10010022 Hydrogen Sulfide 0.2/b 0.2 to 6ppm Pack of 10
12951677 Hydrogen Sulfide 2%/A 2 to 40 vol.% Each
10559722 Hydrogen Sulfide 2/a 2 to 200ppm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15365442 Supplier Dräger™ Supplier No. 6728821
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For gas detection

Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.

Specifications

Components Measured Hydrogen Sulfide 2/a
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 3 to 30mg/L
Measurement Range 2 to 200ppm
No. of Strokes 1/10
Quantity 10
Response Time 3.5 min./20 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
compliance-icons
Product Identifier
  • drager tubes
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contain: hydrogen sulphide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.