missing translation for 'onlineSavingsMsg'
Learn More

SHOWA A0170 Anti-static Knit Gloves

Product Code. 17811201
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17811201 Small 10 pairs
17821201 Medium 10 pairs
17831201 Large 10 pairs
17841201 X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17811201 Supplier SHOWA Supplier No. A017006S
  • ONLINE EXCLUSIVE PRICE
    69.00 €
    55.62 € / 10 pairs
    Save 13.38 €
    19% Off
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The A0170 anti-static glove is suitable for handling glass, abrasive, and electrical components. It has a liner that dissipates static electricity and a polyurethane coating on the palm.

  • The A0170 antistatic glove has a polyurethane coating on the palm that protects against oils and abrasion.
  • The polyurethane coating also leaves no marks or fingerprints on handled objects.
  • The glove is designed for wearer comfort with a flexible, seamless liner.
  • The elasticated cuff of the glove prevents debris from entering.

Specifications

Chlorinated No
Construction Seamless Knit
Accelerator Free Yes
Applications Industrial, Laboratory
Coating Material Polyurethane
Color White, Gray
Disposable Reusable
Exterior Finish Partially Textured
Hand Specific
Laboratory/Cleanroom Use Cleanroom
Latex Free Yes
Material Nylon, Polyester, Polyurethane
Powdered Powder Free
Product Type Anti-static Gloves
Size Small
Sterility Non-sterile
EN388 Standard EN388:3121X
Gender Unisex
Glove Cuff Style Knit
Tear Rating 2
Yarn Gauge 13 G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.