missing translation for 'onlineSavingsMsg'
Learn More

SHOWA A0170 Anti-static Knit Gloves

Product Code. 17841201
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17841201 X-Large 10 pairs
17811201 Small 10 pairs
17821201 Medium 10 pairs
17831201 Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17841201 Supplier SHOWA Supplier No. A017009XL
  • ONLINE EXCLUSIVE PRICE
    69.00 €
    55.62 € / 10 pairs
    Save 13.38 €
    19% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The A0170 anti-static glove is suitable for handling glass, abrasive, and electrical components. It has a liner that dissipates static electricity and a polyurethane coating on the palm.

  • The A0170 antistatic glove has a polyurethane coating on the palm that protects against oils and abrasion.
  • The polyurethane coating also leaves no marks or fingerprints on handled objects.
  • The glove is designed for wearer comfort with a flexible, seamless liner.
  • The elasticated cuff of the glove prevents debris from entering.

Tekniske data

Chlorinated No
Construction Seamless Knit
Accelerator Free Yes
Applications Industrial, Laboratory
Coating Material Polyurethane
Color White, Gray
Disposable Reusable
Exterior Finish Partially Textured
Hand Specific
Laboratory/Cleanroom Use Cleanroom
Latex Free Yes
Material Nylon, Polyester, Polyurethane
Powdered Powder Free
Product Type Anti-static Gloves
Size X-Large
Sterility Non-sterile
EN388 Standard EN388:3121X
Gender Unisex
Glove Cuff Style Knit
Tear Rating 2
Yarn Gauge 13 G
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.