Learn More
SHOWA S-TEX KV3 Cut-Resistant Knit Gloves
Product Code. 17801221
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17801221 Medium 10 pairs
17891211 Small 10 pairs
17811221 Large 10 pairs
17821221 X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17801221 Supplier SHOWA Supplier No. STEX KV308M
  • ONLINE EXCLUSIVE PRICE
    204.00 €
    170.38 € / 10 pairs
    Save 33.62 €
    16% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The S-TEX KV3 glove offers an exceptional level of cut protection while maintaining optimal dexterity.
  • The glove has a natural rubber palm dip that ensures a superior grip in both dry and wet conditions.
  • For maximum cut resistance, the KV3 glove is made with a combination of stainless steel liner, Kevlar fibers, and patented Hagane Coil technology.
  • The KV3 glove is designed to be form-fitting and breathable, ensuring maximum comfort and performance during use.
Specifications
Construction Seamless Knit
Coating Material Latex
Color Black, Yellow
Hand Specific
Latex Free No
Material Aramid Knit, Polyester, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Medium
Gender Unisex
Glove Cuff Style Knit
Puncture Rating ASTM F2878 LEVEL 4