missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ SICCO™ Big Star Desiccator

Product Code. 10420922
Change view
Click to view available options
Volume (Metric):
156 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10420922 156 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10420922 Supplier Bohlender™ Supplier No. V189607
  • ONLINE EXCLUSIVE PRICE
    2694.98 €
    2044.89 € / Pack of 1
    Save 650.09 €
    24% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for storing or drying humidity sensitive products using silica gel. SICCO™ Big Star Desiccators are ideal for storing reference materials, retained samples, metallographic specimen, tobacco and DNA-samples.

  • Tight fitting door with magnetic catch and circular rubber seal protects the contents from contamination
  • Reinforced aluminum frame with acrylic glass panels
  • Non slip rubber feet
  • Easy to read electronic hygrometer
  • Variable height shelves made of stainless steel
  • Telescopic shelves
  • Desiccant tray

Specifications

Color Clear
Material PMMA Acrylic Glass, Aluminum Frame
Product Type Nonvacuum Desiccator
Volume (Metric) 156 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.