Learn More
BOLA SICCO Desiccant
Product Code. 17197714
Change view
Click to view available options
Product Type:
Desiccator Canister
Unit Size:
Each
Product Code. Product Type Unit Size
17197714 Desiccator Canister Each
1 options
Product Code. 17197714 Supplier BOLA Supplier No. V 189508
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-quality silica gel desiccant specifically designed for use in their desiccator cabinets, easy regeneration at approx. 90 - 110°C
  • Silica gel with colour indicator from orange to dark brown and black
  • Does not contain cobalt-II-chloride
  • Grain size 2 - 5 mm, loss after drying less or equal to 4%, Weight 720 g
Specifications
Material Silica Gel
Weight (Metric) 720 g
Product Type Desiccator Canister