missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SID 26681509

Product Code. 18064022
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064022 10 mg 10mg
18044772 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064022 Supplier R&D Systems™ Supplier No. 3625/10
  • ONLINE EXCLUSIVE PRICE
    347.86 €
    311.85 € / 10mg
    Save 36.01 €
    10% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cathepsin L inhibitor

SID 26681509 is a potent and reversible human cathepsin L inhibitor (IC50 = 56 nM). Displays no inhibitory activity at cathepsin G. Antimalarial; inhibits leishmaniasis.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Line Tocris
Product Type SID 26681509
Purity >97%
Molecular Weight (g/mol) 539.65
Inhibitors SID 26681509

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.