Learn More
Berthold Technologies Sideview Kit (IK Version)
Product Code. 31777399 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31777399 Imaging System Each
1 options
Product Code. 31777399 Supplier Berthold Technologies Supplier No. 74647
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
To mount the camera on the side of the imaging chamber in NightSHADE imaging systems.
  • The default camera position in the NightSHADE in vivo imaging system (plant configuration) is on top of the imaging chamber
  • Lateral camera mounting enables side-view imaging
  • Allows seedlings to remain upright in a natural vertical orientation
  • Enables imaging of the entire plant, from leaves to roots
Specifications
Type Connection Kit
For Use With (Application) Plant Imaging
For Use With (Equipment) Imaging System