missing translation for 'onlineSavingsMsg'
Learn More

Tork Signage Holder for Soap and Sanitiser Dispenser

Product Code. 30243806 Shop All Tork Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243806 White Pack of 6
30243801 Black Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30243806 Supplier Tork Supplier No. 511170
  • ONLINE EXCLUSIVE PRICE
    13.47 €
    12.51 € / Pack of 6
    Save 0.96 €
    7% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tork Signage Holder for Tork Soap and Sanitizer Dispensers is a communication tool that encourages good hand hygiene. A perfect way to remind users about the importance of hand hygiene, educate on the correct way to wash their hands or communicate other relevant messages.

  • Promote good hand hygiene with on-dispenser signage for staff and guests.
  • Perfect for reminding users about hand hygiene, showing which refill is in use, educating about the correct way to wash hands, or communicating other relevant messages.
  • Designed to fit S4/S5 Tork soap and sanitiser dispensers.
  • Black or white color creates a seamless and professional appearance when paired with the matching dispenser.
TRUSTED_SUSTAINABILITY

Specifications

Alternate Part Number 7322542116531
Material Plastic
Adhesive Type Non-adhesive
Mounting Type Bracket
For Use With (Application) Promotes Hygiene
Shape Rectangular
Product Type Sign Sleeve
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.