missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ SIL G-25/UV254 Glass Plates

Product Code. 16533860 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 Pc.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16533860 100 Pc. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16533860 Supplier Macherey-Nagel™ Supplier No. 809028.100
  • ONLINE EXCLUSIVE PRICE
    233.00 €
    196.19 € / Pack of 100
    Save 36.81 €
    15% Off
 Restricted Item
In Stock
Add to basket
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Analytical TLC glass plates with unmodified silica layers, stable in organic solvents and featuring a fluorescent indicator for UV detection.

  • SIL G-25 / UV 254 glass plates with unmodified, standard-grade silica 60 layers
  • Analytical plates with unmodified standard silica layers on glass for use in TLC
  • Feature green fluorescent indicator for short-wave UV
  • Manganese-activated zinc silicate indicator with green fluorescence (254 nm)
  • Highly polymeric binders stable in almost all organic solvents
  • Resistant to aggressive visualization reagents
  • Stable in almost all organic solvents

Spezifikation

Backing Material Glass
Material Unmodified Silica Gel (SiOH)
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 100 Pc.
Type Glass Plates Coated with Silica Gel
Fluorescent Indicator Yes
For Use With (Application) Steroids, Antibiotics, Amines, Amino acids, Hormones, Sweeteners, Aflatoxins, Vitamins, Normal Phase (NP), Alkaloids, Analgesics, Marihuana Compounds, Patulin, Standard TLC Applications
For Use With (Equipment) Thin Layer Chromatography (TLC)
Includes 100x TLC Glass Plates (2.5 x 7.5 cm)
Pore Volume 0.75 mL/g
Product Line SIL G
Sorbent SIL G UV254
Specific Surface ∼500 m2/g
Storage Requirements 15°C to 25°C (59°F to 77°F)
Thickness (Metric) 0.25 mm
Dimensions (L x W) 2.5 x 7.5 cm
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.