missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND Automatic Burette with Amber Stopcock

Product Code. 10568021 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10568021 50 mL Each
10132111 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10568021 Supplier BRAND™ Supplier No. 22318
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SILBERBRAND Automatic Burette equipped with an amber lateral stopcock and PTFE key.

  • Error limits according to Class B, DIN EN ISO 385
  • Interchangeable keys
  • Calibrated to deliver (TD, Ex)
  • With glass key in the titration stopcock
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 mL
Graduations 0.1 mL
Includes Without bottle
Stopcock Amber lateral stopcock with PTFE key
Warranty 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.